Learn More
TFIP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14436-1-AP-150UL
 View more versions of this product
Product Code. 16873074
  • £349.00 / 150µL
Description
This gene encodes a protein component of the spliceosome that promotes the release of the lariat-intron during late-stage splicing through the recruitment of a pre-mRNA splicing factor called DEAH-box helicase 15. The encoded protein contains a G-patch domain, a hallmark of RNA-processing proteins, that binds DEAH-box helicase 15. This protein contains an atypical nuclear localization sequence as well as a nuclear speckle-targeting sequence, enabling it to localize to distinct speckled regions within the cell nucleus. Polymorphisms in this gene are associated with dental caries suggesting a role in amelogenesis. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
TFIP11
Polyclonal
Unconjugated
TFIP11
bK445C9.6, STIP, STIP 1, TFIP11, TIP39
Rabbit
Antigen Affinity Chromatography
RUO
24144, 288718, 54723
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5U2Y6, Q9ERA6, Q9UBB9
Tfip11
TFIP11 Fusion Protein Ag5821
150 μL
Primary
Human, Mouse, Rat