missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIP11 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93034-0.1ml
This item is not returnable.
View return policy
Description
TFIP11 Polyclonal antibody specifically detects TFIP11 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TFIP11 | |
| Polyclonal | |
| Western Blot 1:200-1:3000 | |
| bK445C9.6, DKFZP434B194, FLJ22086, hNtr1, NTR1, Septin and tuftelin-interacting protein 1, STIP, STIP-1, TIP39Spp382, tuftelin interacting protein 11, tuftelin-interacting protein 11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human TFIP11 (NP_036275.1). MSLSHLYRDGEGRIDDDDDERENFEITDWDLQNEFNPNRQRHWQTKEEATYGVWAERDSDDERPSFGGKRARDYSAPVNFISAGLKKGAAEEAELEDSDDEEKPVKQDDFPKDFGPRKLKTGGNFKPSQKGFAGGTKSFM | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 24144 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction