missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIIIC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14077
This item is not returnable.
View return policy
Description
TFIIIC Polyclonal specifically detects TFIIIC in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TFIIIC | |
| Polyclonal | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| general transcription factor 3C polypeptide 1, general transcription factor IIIC, polypeptide 1 (alpha subunit), general transcription factor IIIC, polypeptide 1 (alpha subunit, 220kD ), general transcription factor IIIC, polypeptide 1, alpha 220kDa, TF3C-alpha, TFIIIC, TFIIIC 220 kDa subunit, TFIIIC box B-binding subunit, TFIIIC220DKFZp686A111, TFIIICalpha, Transcription factor IIIC 220 kDa subunit, Transcription factor IIIC subunit alpha | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2975 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GTF3C1 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: QVFIFLKKNAVIVDTTICDPHYNLARSSRPFERRLYVLNSMQDVENYWFDLQCVCLNTPLGVVRCPRVRKNSSTDQGSDEEGSLQKEQESAMDKHNLERKCAMLEYTTGSREVVDEGLIPGDGLGAAGLDSSFYG | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction