missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIIE-alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58120
This item is not returnable.
View return policy
Description
TFIIE-alpha Polyclonal specifically detects TFIIE-alpha in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| TFIIE-alpha | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| FE, General transcription factor IIE 56 kDa subunit, general transcription factor IIE subunit 1, general transcription factor IIE, polypeptide 1 (alpha subunit, 56kD), general transcription factor IIE, polypeptide 1, alpha 56kDa, TF2E1, TFIIE-A, TFIIE-alpha, Transcription initiation factor IIE subunit alpha | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| GTF2E1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QSKDHAATTAGAASLAGGHHREAWATKGPSYEDLYTQNVVINMDDQEDLHRASLEGKSAKERPIWLRESTVQGAYGSEDMKEGGIDMDAFQEREEGHAGPDDNEEVMRALLIHEKKTSSAMAGSVGAAAPVTAANGSDSE | |
| 100 μL | |
| Stem Cell Markers | |
| 2960 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction