missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIIB Mouse anti-Human, Clone: PCRP-GTF2B-1D1, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | TFIIB |
|---|---|
| Clone | PCRP-GTF2B-1D1 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
| Applications | Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18342992
|
Novus Biologicals
NBP3-13934-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18304763
|
Novus Biologicals
NBP3-13934-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFIIB Monoclonal antibody specifically detects TFIIB in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Protein ArraySpecifications
| TFIIB | |
| 0.2 mg/ml | |
| Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array | |
| Unconjugated | |
| Mouse | |
| DNA Replication Transcription Translation and Splicing | |
| 10 mM PBS with 0.05% BSA | |
| general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB | |
| Recombinant full-length human TFIIB protein (Uniprot: Q00403) | |
| Primary | |
| Store at 4°C. |
| PCRP-GTF2B-1D1 | |
| Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q00403 | |
| 2959 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title