missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIIB Antibody (PCRP-GTF2B-1D1) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14055JF635
This item is not returnable.
View return policy
Description
TFIIB Monoclonal antibody specifically detects TFIIB in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Protein Array
Specifications
| TFIIB | |
| Monoclonal | |
| Janelia Fluor 635 | |
| general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB | |
| Recombinant full-length human TFIIB protein (Uniprot: Q00403) | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 2959 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunoprecipitation, Protein Array | |
| PCRP-GTF2B-1D1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction