missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TFIIB Monoclonal antibody specifically detects TFIIB in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | TFIIB |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-GTF2B-1D1 |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human TFIIB protein (Uniprot: Q00403) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?