missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIIB Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49981IR
This item is not returnable.
View return policy
Description
TFIIB Polyclonal antibody specifically detects TFIIB in Human samples. It is validated for Western Blot
Specifications
| TFIIB | |
| Polyclonal | |
| Western Blot | |
| general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB | |
| The immunogen for this product maps to a region between residue 100 and 150 of human General Transcription Factor IIB using the numbering given in entry NP_001505.1 (GeneID 2959). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 2959 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction