missing translation for 'onlineSavingsMsg'
Learn More

TFIIB Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18419952 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419952 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419952 Supplier Novus Biologicals Supplier No. NBP149981AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TFIIB Polyclonal antibody specifically detects TFIIB in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFIIB
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 100 and 150 of human General Transcription Factor IIB using the numbering given in entry NP_001505.1 (GeneID 2959).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 2959
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.