missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFG Antibody (TFG-03), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62212H
This item is not returnable.
View return policy
Description
TFG Monoclonal specifically detects TFG in Human samples. It is validated for Western Blot.
Specifications
| TFG | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TFG | |
| Human recombinant protein Trx-his-NTFG | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-57 reacts with gamma-epsilon and delta-epsilon dimers of human CD3 complex, a part of a bigger multisubunit T cell receptor complex (CD3/TCR) expressed on peripheral blood T lymphocytes and mature thymocytes. | |
| Store at 4°C in the dark. |
| Western Blot | |
| TFG-03 | |
| Western Blot | |
| FLJ36137, protein TFG, TF6, TRK-fused gene, TRK-fused gene protein, TRKT3, TRKT3 oncogene | |
| Mouse | |
| Protein A purified | |
| Signal Transduction | |
| 10342 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction