missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TFF2 Monoclonal specifically detects TFF2 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | TFF2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | GE16C |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | SML1trefoil factor 2, SML1, human spasmolytic polypeptide (SP)10spasmolytic protein 1, SP, spasmolysin, Spasmolytic polypeptide, trefoil factor 2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?