missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF2 Mouse anti-Human, Clone:4G7C3, Abnova™
Mouse monoclonal antibody raised against recombinant human TFF2.
Brand: Abnova MAB16649.100ug
This item is not returnable.
View return policy
Description
Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer and affect healing of the epithelium. The encoded protein inhibits gastric acid secretion. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21. [provided by RefSeq]Specifications
| TFF2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| TFF2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 4G7C3 | |
| trefoil factor 2 | |
| In PBS (0.05% sodium azide) | |
| SML1, SP | |
| Recombinant protein corresponding to amino acid 20-125 of human TFF2 from E. coli. | |
| 7032 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction