Learn More
Invitrogen™ TFF1 Recombinant Rabbit Monoclonal Antibody (HL2968)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556895
Product Code. 30276426
  • £434.00 / 100µL
Description
TFF1 Recombinant Monoclonal Antibody for Western Blot
Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TFF1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P04155
TFF1
Full length human TFF1 recombinant protein.
100 μL
Primary
Human
Antibody
IgG
Western Blot
HL2968
Unconjugated
TFF1
Bcei; breast cancer estrogen-inducible protein; breast cancer estrogen-inducible sequence; D21S21; gastrointestinal trefoil protein pS2; HP1.A; HPS2; PNR-2; polypeptide P1.A; Protein pS2; PS2; TFF1; Trefoil factor 1
Rabbit
Protein A
RUO
7031
-20°C, Avoid Freeze/Thaw Cycles