missing translation for 'onlineSavingsMsg'
Learn More

TFF1/pS2 Mouse anti-Human, Clone: TFF1/2133, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07341-100ug

 View more versions of this product

Product Code. 18674309

  • £377.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TFF1/pS2 Monoclonal specifically detects TFF1/pS2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TFF1/pS2
Monoclonal
0.2 mg/mL
Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Protein Array
BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A
Recombinant full-length human TFF1/pS2 protein
100 μg
Breast Cancer, Cancer
7031
Store at 4°C.
IgG2b κ
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array
TFF1/2133
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.