missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF1/pS2 Antibody (PS2GE2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £302.00
Specifications
| Antigen | TFF1/pS2 |
|---|---|
| Clone | PS2GE2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 0.5 - 1.0 μg/106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 0.5 - 1 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 0.5 - 1 μg/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617467
|
Novus Biologicals
NBP2-50275-0.025ml |
0.025 mL |
£159.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667916
|
Novus Biologicals
NBP2-50275 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFF1/pS2 Monoclonal antibody specifically detects TFF1/pS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOFSpecifications
| TFF1/pS2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer | |
| PBS | |
| 7031 | |
| IgG1 κ | |
| Protein A purified |
| PS2GE2 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 0.5 - 1.0 μg/106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 0.5 - 1 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 0.5 - 1 μg/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A | |
| A synthetic peptide of 28 amino acid residues from the C-terminus of human TFF1/pS2 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title