missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFEC Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a member of the micropthalmia (MiT) family of basic helix-loop-helix leucine zipper transcription factors. MiT transcription factors regulate the expression of target genes by binding to E-box recognition sequences as homo- or heterodimers, and play roles in multiple cellular processes including survival, growth and differentiation. The encoded protein is a transcriptional activator of the nonmuscle myosin II heavy chain-A gene, and may also co-regulate target genes in osteoclasts as a heterodimer with micropthalmia-associated transcription factor. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
Specifications
| Antigen | TFEC |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Tfec |
| Gene Accession No. | O14948 |
| Gene Alias | BB107417; bHLHe34; Class E basic helix-loop-helix protein 34; hTFEC-L; mTFEC; rTFEC; TCFEC; TFEC; TFE-C; TFECL; TFEC-L; Transcription factor EC; Transcription factor EC-like |
| Gene Symbols | Tfec |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?