missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TFEB Monoclonal Antibody (CL14112)

Product Code. 30830029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30830029 Supplier Invitrogen™ Supplier No. MA563039
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors. Recent studies have shown that lysosomal biogenesis is regulated by TFEB, which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1. Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFEB
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL14112
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TFEB
Gene Accession No. P19484
Gene Alias Alpha TCFEB; ALPHATFEB; BHLHE35; Class E basic helix-loop-helix protein 35; RP4-696P19.3; T-cell transcription factor EB; Tcfeb; Tfeb; Transcription factor EB
Gene Symbols TFEB
Host Species Mouse
Immunogen Recombinant protein corresponding to Human TFEB.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7942
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.