missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEB Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41167AF532
This item is not returnable.
View return policy
Description
TFEB Polyclonal antibody specifically detects TFEB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TFEB | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human TFEB. The immunogen is located within amino acids 70 - 120 of TFEB. Amino Acid Squence: SQHQKVREYLSETYGNKF | |
| 0.1 mL | |
| Autophagy | |
| 7942 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction