missing translation for 'onlineSavingsMsg'
Learn More

TFDP3 Antibody (3F11), Novus Biologicals™

Product Code. 18366689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366689 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366689 Supplier Novus Biologicals Supplier No. H00051270M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TFDP3 Monoclonal antibody specifically detects TFDP3 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFDP3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3F11
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_057605
Gene Alias CT30MGC161639, DP4, E2F-like, E2F-like protein, HCA661Cancer/testis antigen 30, Hepatocellular carcinoma-associated antigen 661, transcription factor Dp family member 3, transcription factor Dp family, member 3
Host Species Mouse
Immunogen TFDP3 (NP_057605, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPQRPAASNIPVVGSPNPPSTHFASQNQHSYSSPPWAGQHNRKGEKNGMGLCRLSMKVWETVQRKGTTSCQEVVGELVAKFRAASNHASPNESAYDVKNI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51270
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.