missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFDP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11500-1-AP-150UL
This item is not returnable.
View return policy
Description
The gene is a member of the transcription factor DP family. The encoded protein forms heterodimers with the E2F transcription factors resulting in transcriptional activation of cell cycle regulated genes. Alternative splicing results in multiple transcript variants.Specifications
| TFDP2 | |
| Polyclonal | |
| Unconjugated | |
| TFDP2 | |
| Dp 2, DP2, E2F dimerization partner 2, TFDP2, Transcription factor Dp 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 211586, 300947, 7029 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14188, Q64163 | |
| TFDP2 | |
| TFDP2 Fusion Protein Ag2054 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction