missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFCP2L1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | TFCP2L1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TFCP2L1 Polyclonal specifically detects TFCP2L1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| TFCP2L1 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Transcription Factors and Regulators | |
| PBS buffer, 2% sucrose | |
| 29842 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml, Immunohistochemistry | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CRTR1CRTR-1, LBP-9, LBP9CP2-related transcriptional repressor 1, transcription factor CP2-like 1, transcription factor CP2-like protein 1, Transcription factor LBP-9 | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human TFCP2L1 (NP_055368). Peptide sequence LRDVLALPIFKQEEPQLSPENEARLPPLQYVLCAATSPAVKLHEETLTYL | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title