missing translation for 'onlineSavingsMsg'
Learn More

TFCP2L1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93849-0.1ml

 View more versions of this product

Product Code. 18653111

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

TFCP2L1 Polyclonal antibody specifically detects TFCP2L1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TFCP2L1
Polyclonal
Western Blot 1:500-1:2000
CRTR1CRTR-1, LBP-9, LBP9CP2-related transcriptional repressor 1, transcription factor CP2-like 1, transcription factor CP2-like protein 1, Transcription factor LBP-9
Recombinant fusion protein containing a sequence corresponding to amino acids 360-430 of human TFCP2L1 (NP_055368.1). NAIKGRNVRPKMTIYVCQELEQNRVPLQQKRDGSGDSNLSVYHAIFLEELTTLELIEKIANLYSISPQHIH
0.1 mL
Transcription Factors and Regulators
29842
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.