missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFCP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15203-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a transcription factor that binds the alpha-globin promoter and activates transcription of the alpha-globin gene. The encoded protein regulates erythroid gene expression, plays a role in the transcriptional switch of globin gene promoters, and it activates many other cellular and viral gene promoters. The gene product interacts with certain inflammatory response factors, and polymorphisms of this gene may be involved in the pathogenesis of Alzheimer′s disease.Specifications
| TFCP2 | |
| Polyclonal | |
| Unconjugated | |
| TFCP2 | |
| CP2, LBP 1C, LSF, SAA3 enhancer factor, SEF, TFCP2, TFCP2C, transcription factor CP2, Transcription factor LSF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21422, 315309, 7024 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q12800, Q9ERA0 | |
| Tfcp2 | |
| TFCP2 Fusion Protein Ag7353 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction