missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFB1M Polyclonal Antibody

TFB1M Polyclonal Antibody
Brand: Invitrogen PA5110477
This item is not returnable.
View return policy
Description
TFB1M Polyclonal Antibody for Western Blot
Specifications
| TFB1M | |
| Polyclonal | |
| Unconjugated | |
| TFB1M | |
| CGI75; CGI-75; Dimethyladenosine transferase 1, mitochondrial; h-mtTFB; h-mtTFB1; homolog of yeast mitochondrial transcription factor B; hTFB1M; mitochondrial 12S rRNA dimethylase 1; mitochondrial dimethyladenosine transferase 1; Mitochondrial transcription factor B1; mtTFB; mtTFB1; OTTHUMP00000017473; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 1; TFB1M; transcription factor B1, mitochondrial | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 224481, 308140, 51106 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q811P6, Q8JZM0, Q8WVM0 | |
| TFB1M | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-346 of human TFB1M (NP_057104.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction