missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFAP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595836
This item is not returnable.
View return policy
Description
Cellular Location: Nucleus Immunogen sequence: QQEQVRLLHQ EKLEREQQQL RTQLLPPPAP THHPTVIVPA PPPPPSHHIN VVTMGPSSVI NSVSTSRQNL DTIVQAIQHI EGTQEKQELE EEQRRAVIVK PVRSCPEAPT SDTASDSEAS DSDAMDQSRE EPSGDGELP.
Transcription factors of the basic helix-loop-helix-zipper(bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization. Transcription factor AP4 activates both viral and cellular genes by binding to the symmetrical DNA sequence CAGCTG.
Specifications
| TFAP4 | |
| Polyclonal | |
| Unconjugated | |
| Tfap4 | |
| Activating enhancer-binding protein 4; AI642933; AP4; AP-4; bHLHc41; Class C basic helix-loop-helix protein 41; D930048N17Rik; LOW QUALITY PROTEIN: transcription factor AP-4; Tcfap4; TFAP4; transcription factor AP4; Transcription factor AP-4; transcription factor AP-4 (activating enhancer binding protein 4); transcription factor AP-4 (activating enhancer-binding protein 4) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 360482, 7023 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.6 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q01664 | |
| Tfap4 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-338 of human TFAP4 (NP_003214.1). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction