missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAP2D Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-47200
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TFAP2D Polyclonal specifically detects TFAP2D in Human, Mouse samples. It is validated for Peptide ELISA.
Spécification
| TFAP2D | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Activating enhancer-binding protein 2-delta, AP-2 like, AP2-delta, TFAP2BL1activating enhancer binding protein 2 beta-like 1, transcription factor AP-2 beta (activating enhancer binding protein 2beta)-like 1, transcription factor AP-2 beta (activating enhancer-binding protein 2beta)-like 1, transcription factor AP-2 delta (activating enhancer binding protein 2 delta), Transcription factor AP-2-beta-like 1, transcription factor AP-2-delta | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 83741 | |
| Human, Mouse | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:2000 | |
| NP_758438.2 | |
| TFAP2D | |
| Peptide with sequence C-HHQSFHYEFQHSH corresponding to internal region according to NP_758438.2. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction