missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAP2C Mouse anti-Human, Clone:2F12A6, Abnova™
Mouse monoclonal antibody raised against partial recombinant human TFAP2C.
Brand: Abnova MAB17908.100ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a sequence-specific DNA-binding transcription factor involved in the activation of several developmental genes. The encoded protein can act as either a homodimer or heterodimer with other family members and is induced during retinoic acid-mediated differentiation. It plays a role in the development of the eyes, face, body wall, limbs, and neural tube. [provided by RefSeq]Specifications
| TFAP2C | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| TFAP2C | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 2F12A6 | |
| transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma) | |
| In PBS (0.05% sodium azide) | |
| AP2-GAMMA, ERF1, TFAP2G, hAP-2g | |
| Recombinant protein corresponding to amino acids 341-450 of human TFAP2C. | |
| 7022 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction