missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAP2A, Mouse anti-Human, Clone: 2G5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007020-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: SGLLHTHRGLPHQLSGLDPRRDYRRHEDLLHGPHALSSGLGDLSIHSLPHAIEEVPHVEDPGINIPDQTVIKKGPVSLSKSNSNAVSAIPINKDNLFGGVVNPNEVFSpecifications
| TFAP2A | |
| Monoclonal | |
| Unconjugated | |
| transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha) | |
| AP-2/AP-2alpha/AP2TF/BOFS/TFAP2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), KnockDown, Western Blot | |
| 2G5 | |
| In 1x PBS, pH 7.4 | |
| BC017754 | |
| TFAP2A | |
| TFAP2A (AAH17754, 99 a.a. ∼ 205 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7020 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction