missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFAM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589298
This item is not returnable.
View return policy
Description
Immunogen sequence: QDAYRAEWQV YKEEISRFKE QLTPSQIMSL EKEIMDKHLK RKAMTKKKEL TLLGKPKRPR SAYNVYVAER FQEAKGDSPQ EKLKTVKENW KNLSDSEKEL Y Positive Samples: SW620, HT-29; Cellular Location: Mitochondrion, Mitochondrion matrix, mitochondrion nucleoid.
Mitochondrial transcription factor A (TFAM) is a crucial protein encoded by nuclear genes, but it is transported from the cytoplasm to mitochondria where it plays a key role in the maintenance and expression of mitochondrial DNA (mtDNA). It regulates the replication and transcription of mtDNA, ensuring the proper functioning of mitochondrial processes. Structurally, TFAM contains a molecular region known as the LC3 interacting region (LIR) motif, which allows it to bind to autophagy proteins such as LC3. This interaction is essential for the autolysosomal pathway, helping to eliminate leaked mtDNA and limit inflammation. TFAM levels vary across tissues, with notable effects on mitochondrial function; high levels can repress mtDNA expression in certain tissues, such as skeletal muscle, and contribute to deficiencies in oxidative phosphorylation (OXPHOS). Conversely, in other tissues like the heart, increased mtDNA copy number maintains a balanced TFAM-to-mtDNA ratio, preserving OXPHOS capacity. TFAM is integral to the packaging, stability, and replication of the mitochondrial genome, and disruptions in TFAM can have severe effects, including implications in neurodegeneration and other diseases related to mtDNA stress.
Specifications
| TFAM | |
| Polyclonal | |
| Unconjugated | |
| Tfam | |
| AI661103; Hmgts; Mitochondrial transcription factor 1; mitochondrial transcription factor A; MTTF1; mtTFA; TCF6; TCF-6; TCF6L1; TCF6L2; TCF6L3; Testis-specific high mobility group protein; testis-specific HMG-box protein m-tsHMG; TFAM; Transcription factor 6; transcription factor 6-like 1; transcription factor 6-like 2; transcription factor 6-like 2 (mitochondrial transcription factor); transcription factor 6-like 3; transcription factor A, mitochondrial; tsHMG; TS-HMG | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21780, 7019, 83474 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 7.83 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P40630, Q00059, Q91ZW1 | |
| Tfam | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction