Learn More
Invitrogen™ TFAM Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA580106
Product Code. 15905675
  • £400.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human K562 whole cell, human CACO-2 whole cell, huamn Raji whole cell. IHC: human breast cancer tissue, human intestinal diffuse large B-cell lymphoma tissue, human ovarian serous adenocarcinoma tissue, human tonsil tissue, human liver cancer tissue, human lung adenocarcinoma tissue, human colorectal adenocarcinoma tissue. ICC/IF: U2OS cell.
Mitochondrial transcription factor A (TFAM) is a crucial protein encoded by nuclear genes, but it is transported from the cytoplasm to mitochondria where it plays a key role in the maintenance and expression of mitochondrial DNA (mtDNA). It regulates the replication and transcription of mtDNA, ensuring the proper functioning of mitochondrial processes. Structurally, TFAM contains a molecular region known as the LC3 interacting region (LIR) motif, which allows it to bind to autophagy proteins such as LC3. This interaction is essential for the autolysosomal pathway, helping to eliminate leaked mtDNA and limit inflammation. TFAM levels vary across tissues, with notable effects on mitochondrial function; high levels can repress mtDNA expression in certain tissues, such as skeletal muscle, and contribute to deficiencies in oxidative phosphorylation (OXPHOS). Conversely, in other tissues like the heart, increased mtDNA copy number maintains a balanced TFAM-to-mtDNA ratio, preserving OXPHOS capacity. TFAM is integral to the packaging, stability, and replication of the mitochondrial genome, and disruptions in TFAM can have severe effects, including implications in neurodegeneration and other diseases related to mtDNA stress.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TFAM
Polyclonal
Unconjugated
Tfam
AI661103; Hmgts; Mitochondrial transcription factor 1; mitochondrial transcription factor A; MTTF1; mtTFA; TCF6; TCF-6; TCF6L1; TCF6L2; TCF6L3; Testis-specific high mobility group protein; testis-specific HMG-box protein m-tsHMG; TFAM; Transcription factor 6; transcription factor 6-like 1; transcription factor 6-like 2; transcription factor 6-like 2 (mitochondrial transcription factor); transcription factor 6-like 3; transcription factor A, mitochondrial; tsHMG; TS-HMG
Rabbit
Antigen affinity chromatography
RUO
7019
-20°C
Lyophilized
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
500 μg/mL
PBS with 4mg trehalose and no preservative
Q00059
Tfam
A synthetic peptide corresponding to a sequence at the C-terminus of human mtTFA (227-246aa RKDLLRRTIKKQRKYGAEEC).