Learn More
Texwipe™ Replacement Covers for AlphaMop™ Cleanroom Mops
Shop All Texwipe Products
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
1 set
Description
Includes
Lot-specific certificates of compliance and irradiation
- Made of pre-sterilized double-knit AlphaWipe™ polyester
- Gamma irradiated; covers and pads double-bagged together for sterlity assurance
- For use in controlled, aseptic and sterile environments
- Mopping walls, floors and ceilings
- Applying and removing cleaning and disinfecting solutions
- Extremely low levels of releasable fibers
- Featured fitting compatibility with lat-padded head mop easily changeable, laundered polyester
- Completely independent quality control audits for sterility assurance
- Easy-to-read lot code and expiration date on each bag
- Eliminates preparation, documentation and validation of autoclaving covers and pads
- Conforms to textured floors and other surfaces
Compliance
Sterility Assurance Level (SAL) of 106 in accordance with ANSI™/AAMI™/ISO™11137
Specifications
Specifications
| Autoclavable | Autoclavable |