Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 30 Layer
Apply directly to floor
Brand:  Texwipe™ AMA254581G
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15762386
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Apply directly to floor
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Specifications
25 x 45 in.
Gray
30 Sheets/Mat