Learn More
Texwipe™ CleanTip™ Swabs: High-Sorbency Series
Certified to be silicone free
Brand: Texwipe™ TX700B*
Code : NEW
Additional Details : Weight : 1.65600kg
Description
High Sorbency Series
Highly sorptive swabs for industrial applications such as cleaning with solvents such as IPA, applying lubricants and other liquids, and removing excess materials.
- Cotton bud head available with or without an open-cell polyurethane foam sheath
- Soft, nonabrasive foam cover protects against fiber release
- Long handles constructed of hardwood for assured strength and rigidity
- Constructed from the cleanest materials available and manufactured in a cleanroom using state-of-the-art processes and equipment
- High-precision automated process guarantees consistent performance, sizing and cleanliness
- Silicone-free Bag-Within-A-Bag™ packaging consists of a solvent-resistant, thermally sealed exterior bag that protects the inner bags containing the swabs
- Each outer bag is individually lot-coded for easy traceability and quality control