Learn More
Description
TexTra™ Non-sterile Dry Cleanroom Wipers are made from 100% polyester with a cut edge. These wipers are made from thick, cleanroom polyester knit. These single-ply wipers have the liquid-holding capacity of two-ply wipers. TexTra™ combines this high absorbency with great abrasion resistance, resulting in a wiper that is great for wiping and cleaning surfaces, equipment, and parts. Furthermore, it is suitable in applying and removing lubricants, adhesives, residues, and other solutions including disinfectants.
- High absorbency wiper ideal for spill control, cleaning, and solution application
- Cleanroom laundered for low levels of ions and NVRs (non-volatile residues) particles
- Excellent chemical resistance for compatibility with a variety of solutions
- Meets USP 797 and USP 800 wiper requirements
Specifications
Specifications
| Material | Polyester |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO Class 3 to 8, Class 1 to 100,000 |
| Color | White |
| Edges | Cut |
| For Use With (Application) | Cleaning and Solution Application |
| Sterility | Non-sterile |
| Certifications/Compliance (Europe) | Grade A to D |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.