Description
- Formulated for critical cleanroom use, providing exceptional contamination control characteristics.
- Ensures very low particle generation and ion levels for clean and controlled performance.
- High opacity rating enhances readability and print clarity in all applications.
- Precision-cut edges ensure smooth feeding and prevent particulate shedding during printing.
- Cleanroom packaged to maintain product integrity and prevent external contamination.
- Delivers outstanding copying and writing quality with clear, consistent reproductions.
- Compatible with laser printers, photocopiers, inkjet, and offset printing processes.
- Autoclavable for use in sterile environments requiring contamination-free documentation.
- Ideal for producing cleanroom manuals, work instructions, note-taking, data transfer sheets, and color-coded carriers.
- Available in multiple color options to support document categorization in controlled environments.
Specifications
Specifications
| Material | Latex-impregnated Paper |
| Ruled | No |
| Color | Orange |
| Cover Color | Clear |
| Dimensions (L x W) | 210 x 297 mm |
| Autoclavable | Not Autoclavable |
| Disposable | Single-use |
| Product Type | Cleanroom Bond Paper |
| Sterility | Non-sterile |