missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ Texas Red/mCherry/AlexaFluor 594 Set
Click to view available options
For Use With (Equipment):
BA410E, and AE31E series
Unit Size:
Each
Description
- Fluorescence filter set/cube
- Exciter
- Dichroic
- Emitter
- Suitable for the following:
- BA410E Binocular Sextuple 50 W
- BA410E Binocular Sextuple 100 W
- BA410E Trinocular Sextuple 50 W
- BA10E Trinocular Sextuple 100 W
- AE31E Binocular
- AE31E Trinocular
- AE31E Binocular 100 W
- AE31E Trinocular 100 W
Specifications
Specifications
| For Use With (Equipment) | BA410E, and AE31E series |
| Includes | AT560/40x Exciter, AT600DC Dichroic, AT635/60 m Emitter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction