missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TEX55 Polyclonal specifically detects TEX55 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | TEX55 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | uncharacterized protein C3orf30, chromosome 3 open reading frame 30 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human TEX55 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?