missing translation for 'onlineSavingsMsg'
Learn More

TEX30 Antibody (OTI7C3), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-44910-100ul

 View more versions of this product

Product Code. 30306397

  • £417.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TEX30 Monoclonal antibody specifically detects TEX30 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TEX30
Monoclonal
Unconjugated
PBS (pH 7.3) 1% BSA, 50% glycerol
Mouse
>95%
RUO
93081
Store at -20 degrees C. Avoid freeze/thaw cycles.
Purified
Western Blot
OTI7C3
Western Blot 1:500
C13orf27, chromosome 13 open reading frame 27, hypothetical protein LOC93081, RP11-484I6.3, testis expressed 30
Full length human recombinant protein of human TEX30(NP_620134) produced in E.coli.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.