missing translation for 'onlineSavingsMsg'
Learn More

TEX30 Antibody (OTI7C3) [DyLight 488], Novus Biologicals™

Product Code. 30687395 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687395 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687395 Supplier Novus Biologicals Supplier No. NBP344910G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TEX30 Monoclonal antibody specifically detects TEX30 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX30
Applications Western Blot
Classification Monoclonal
Clone OTI7C3
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias C13orf27, chromosome 13 open reading frame 27, hypothetical protein LOC93081, RP11-484I6.3, testis expressed 30
Host Species Mouse
Immunogen Full length human recombinant protein of human TEX30(NP_620134) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93081
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.