missing translation for 'onlineSavingsMsg'
Learn More

TEX28 Antibody (1A8), Novus Biologicals™

Product Code. 18398677 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398677 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398677 Supplier Novus Biologicals Supplier No. H00001527M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TEX28 Monoclonal antibody specifically detects TEX28 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX28
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH26183
Gene Alias chromosome X open reading frame 2, CXorf2, fTEX, testis expressed 28, testis-specific protein TEX28
Host Species Mouse
Immunogen TEX28 (AAH26183, 1 a.a. ~ 410 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVLKAEHTRSPSATLPSNVPSCRSLSSSEDGPSGPSSLADGGLAHNLQDSVRHRILYLSEQLRVEKASRDGNTVSYLKLVSKADRHQVPHIQQAFEKVNQRASATIAQIEHRLHQCHQQLQELEEGCRPEGLLLMAESDPANCEPPSEKALLSEPPEPGGEDGPVNLPHASRPFILESRFQSLQQGTCLETEDVAQQQNLLLQKVKAELEEAKRFHISLQESYHSLKERSLTDLQLLLESLQEEKCRQALMEEQVNGRLQGQLNEIYNLKHNLACSEERMAYLSYERAKEIWEITETFKSRISKLEMLQQVTQLEAAEHLQSRPPQMLFKFLSPRLSLATVLLVFVSTLCACPSSLISSRLCTCTMLMLIGLGVLAWQRWRAIPATDWQEWVPSRCRLYSKDSGPPADGP
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1527
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.