missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEX264 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25858-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| TEX264 | |
| Polyclonal | |
| Unconjugated | |
| TEX264 | |
| SIG11, testis expressed 264, TEX264, ZSIG11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21767, 51368 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y6I9 | |
| TEX264 | |
| TEX264 Fusion Protein Ag23027 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction