missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEX22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564229
This item is not returnable.
View return policy
Description
Immunogen sequence: QLHCRDVVQM VAQLVSEDVD KDVLLPHPLR STESTNAFQA FLARSAPFWH NATFEAS Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 47%.
Specifications
| TEX22 | |
| Polyclonal | |
| Unconjugated | |
| TEX22 | |
| testis expressed 22; testis expressed gene 22; Testis-expressed protein 22; testis-expressed sequence 22 protein; TEX22 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 647310 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| C9J3V5 | |
| TEX22 | |
| Recombinant protein corresponding to Human TEX22. Recombinant protein control fragment (Product #RP-103603). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction