missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TEX2 Antibody, Novus Biologicals™

Product Code. 18347129 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347129 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347129 Supplier Novus Biologicals™ Supplier No. H00055852B01P

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Polyclonal Antibody

TEX2 Polyclonal antibody specifically detects TEX2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen TEX2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH40521.1
Gene Alias HT008, KIAA1738DKFZp781G0721, testis expressed 2, testis expressed sequence 2, testis-expressed sequence 2 protein, TMEM96, transmembrane protein 96
Host Species Mouse
Immunogen TEX2 (AAH40521.1, 1 a.a. - 1134 a.a.) full-length human protein. MTSLYGRHAEKTTDMPKPSAPKVHVQRSVSRDTIAIHFSASGEEEEEEEEEFKEYFEEGLDDQSIVTGLEAKEDLYLEPQVGHDPAGPAASPVLADGLSVSQAPAILPVSKNTVKLLESPVPAAQVLSTVPLAVSPGSSSSGPLASSPSVSSLSEQKTSSSSPLSSPSKSPILSSSASTSTLSSAKPFMSLVKSLSTEVEPKESPHPARHRHLMKTLVKSLSTDTSRQESDTVSYKPPDSKLNLHLFKQFTQPRNTGGDSKTAPSSPLTSPSDTRSFFKVPEMEAKIEDTKRRLSEVIYEPFQLLSKIIGEESGSHRPKALSSSASELSNLSSLNGHLESNNNYSIKEEECDSEGDGYGSDSNIPRSDHPKSTGEPTREIELKSSQGSSLKDLGLKTSSLVLEKCSLSALVSKEDEEFCELYTEDFDLETEGESKVDKLSDIPLKPEVLAKDGVVLDSEDEVDSAVQHPELPVKTLGFFIMCVYVYLILPLPHYVSGLFLGIGLGFMTAVCVIWFFTPPSAHKYHKLHKNLRHWNTRSLDIKEPEILKGWMNEIYNYDPETYHATLTHSVFVRLEGGTLRLSKPNKNISRRASYNEPKPEVTYISQKIYDLSDSKIYLVPKTLARKRIWNKKYPICIELGQQDDFMSKAQTDKETSEEKPPAEGSEDPKKPPRPQEGTRSSQRDQILYLFGRTGREKEEWFRRFILASKLKSEIKKSSGVSGGKPAPVFLAPGLLPAHSRHNSPSGHLTHSRSSSKGSVEEIMSQPKQKELAGSVRQKMLLDYSVYMGRCVPQESRSPQRSPLQSAESSPTAGKKLPEVPPSEEEEQEAWVNALLGRIFWDFLGEKYWSDLVSKKIQMKLSKIKLPYFMNELTLTELDMGVAVPKILQAFKPYVDHQGLWIDLEMSYNGSFLMTLETKMNLTKLGKEPLVEALKVGEIGKEGCRPRAFCLADSDEESSSAGSSEEDDAPEPSGGDKQLLPGAEGYVGGHRTSKIMRFVDKITKSKYFQKATETEFIKKKIEEVSNTPLLLTVEVQECRGTLAVNIPPPPTDRVWYGFRKPPHVELKARPKLGEREVTLVHVTDWIEKKLEQEFQKVFVMPNMDDVYITIMHSAMDPRSTSCLLKDPPVEAADQP
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 55852
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.