missing translation for 'onlineSavingsMsg'
Learn More

Tex14 Antibody (4E4), Novus Biologicals™

Product Code. 18387929 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387929 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387929 Supplier Novus Biologicals Supplier No. H00056155M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tex14 Monoclonal antibody specifically detects Tex14 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tex14
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4E4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_938207.2
Gene Alias cancer/testis antigen 113, CT113, Protein kinase-like protein SgK307, SGK307, Sugen kinase 307, testis expressed 14, testis expressed sequence 14, testis-expressed protein 14, Testis-expressed sequence 14
Host Species Mouse
Immunogen TEX14 (NP_938207.2, 1392 a.a. ~ 1491 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DTERAHSTLDEDLERWLQPPEESVELQDLPKGSERETNIKDQKVGEEKRKREDSITPERRKSEGVLGTSEEDELKSCFWKRLGWSESSRIIVLDQSDLSD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56155
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.