missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TEX11 Recombinant Mouse Monoclonal Antibody (K94052_9C7)
GREENER_CHOICE

Product Code. 30827095
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827095 100 μL
30827413 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827095 Supplier Invitrogen™ Supplier No. MA564457

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: LQAIITLENI LTDEESEDND LVAERGSPTM LLSLAAQFAL ENGQQIVAEK ALEYLAQHSE DQEQVLTAVK CLLRFLLPKI AEMPESEDKK KEMDRLLTCL NRAFVKLSQP Highest antigen sequence identity to the following orthologs: Mouse - 53%, Pig - 49%, Cynomolgus monkey - 96%.

This gene is X-linked and is expressed in only male germ cells. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX11
Applications Immunoprecipitation
Classification Recombinant Monoclonal
Clone K94052_9C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene TEX11
Gene Accession No. Q8IYF3
Gene Alias 4930565P14Rik; BB024231; Protein ZIP4 homolog; RGD1560239; SPGFX2; testis expressed 11; testis expressed gene 11; testis expressed sequence 11; testis-expressed protein 11; testis-expressed sequence 11 protein; testis-expressed sequence 11 protein; testis-expressed protein 11; Tex11; TGC1; TSGA3; ZIP4H
Gene Symbols TEX11
Host Species Mouse
Immunogen Recombinant human TEX11 (aa 511-620).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56159
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.