missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEX11 Recombinant Mouse Monoclonal Antibody (K94052_6F11)
Description
Immunogen sequence: LQAIITLENI LTDEESEDND LVAERGSPTM LLSLAAQFAL ENGQQIVAEK ALEYLAQHSE DQEQVLTAVK CLLRFLLPKI AEMPESEDKK KEMDRLLTCL NRAFVKLSQP Highest antigen sequence identity to the following orthologs: Mouse - 53%, Pig - 49%, Cynomolgus monkey - 96%.
This gene is X-linked and is expressed in only male germ cells. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
Specifications
| Antigen | TEX11 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Recombinant Monoclonal |
| Clone | K94052_6F11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | TEX11 |
| Gene Accession No. | Q8IYF3 |
| Gene Alias | 4930565P14Rik; BB024231; Protein ZIP4 homolog; RGD1560239; SPGFX2; testis expressed 11; testis expressed gene 11; testis expressed sequence 11; testis-expressed protein 11; testis-expressed sequence 11 protein; testis-expressed sequence 11 protein; testis-expressed protein 11; Tex11; TGC1; TSGA3; ZIP4H |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?