missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEX101 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15447-1-AP-20UL
This item is not returnable.
View return policy
Description
May play a role in signal transduction and promote protein tyrosine phosphorylation.Specifications
| TEX101 | |
| Polyclonal | |
| Unconjugated | |
| TEX101 | |
| Cell surface receptor NYD SP8, GTPR867, NYD SP8, PRO1884, SGRG, TES101RP, testis expressed 101, Testis expressed protein 101, TEX101 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83639 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BY14 | |
| TEX101 | |
| TEX101 Fusion Protein Ag7611 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction