missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TEX101 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA559784

Product Code. 15777631

  • £444.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ELYCQKGLSM TVEADPANMF NWTTEEVETC DKGALCQETI LIIKAGTETA ILATKGCIPE GEEAITIVQH SSPPG Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 49%.

May play a role in signal transduction and promote protein tyrosine phosphorylation. Target protein is a glycoprotein whose aggregation induced a rapid increase in tyrosine phosphorylation of Syk kinase and LAT adaptor, calcium flux, and release of secretory components.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TEX101
Polyclonal
Unconjugated
TEX101
cancer/testis antigen 131; cell surface receptor NYD-SP8; CT131; GTPR867; NYD-SP8; PRO1884; Scleroderma-associated autoantigen; SGRG; SPATA44; spermatogenesis associated 44; spermatogenesis-related gene protein; TES101RP; testis expressed 101; testis expressed sequence 101; testis-expressed protein 101; testis-expressed sequence 101 protein; testis-specific protein TES101RP; TEX101; UNQ867/PRO1884
Rabbit
Antigen affinity chromatography
RUO
83639
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BY14
TEX101
Recombinant protein corresponding to Human TEX101. Recombinant protein control fragment (Product #RP-99089).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.