missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEX101, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00083639-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MGAMQIQTSSSQTSPEEAMGTPRIQHLLILLVLGASLLTSGLELYCQKGLSMTVEADPANMFNWTTEEVETCDKGALCQETILIIKAGTETAILATKGCIPEGEEAITIVQHSSPPGLIVTSYSNYCEDSFCNDKDSLSQFWEFSETTASTVSTTLHCPTCVALGTCFSAPSLPCPNGTTRCYQGKLEITGGGIESSVEVKGCTAMIGCRLMSGILAVGPMFVREACPHQLLTQPRKTENGATCLPIPVWGLQLLLPLLLPSFIHFSSpecifications
| TEX101 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC001861 | |
| TEX101 | |
| TEX101 (AAH01861, 1 a.a. ∼ 267 a.a) full-length human protein. | |
| RUO | |
| 83639 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| testis expressed 101 | |
| GTPR867/MGC4766/NYD-SP8/PRO1884/SGRG | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction