missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEX101 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-88430
This item is not returnable.
View return policy
Description
TEX101 Polyclonal specifically detects TEX101 in Human samples. It is validated for Western Blot.
Specifications
| TEX101 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 83639 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| cancer/testis antigen 131, Cell surface receptor NYD-SP8, CT131, MGC4766, NYD-SP8, PRO1884, Scleroderma-associated autoantigen, SGRGGTPR867, Spermatogenesis-related gene protein, TES101RP, testis expressed 101, testis expressed sequence 101, testis-expressed protein 101, testis-specific protein TES101RP | |
| The immunogen is a synthetic peptide directed towards the middle region of Human TEX101 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction